4'-tert-Butyl-4-chlorobutyrophenone ≥95.0% (by GC)

Supplier: TCI America

Synonyms: 1-(4-(tert-Butyl)phenyl)-4-chlorobutan-1-one

B3144-25G
TCB3144-25GEA 76 USD
TCB3144-25G
4'-tert-Butyl-4-chlorobutyrophenone ≥95.0% (by GC)
4'-tert-Butyl-4-chlorobutyrophenone

CAS Number: 43076-61-5
MDL Number: MFCD00018996
Molecular Formula: C14H19ClO
Molecular Weight: 238.76
Purity/Analysis Method: 95.0% (GC)
Form: Crystal
Boiling point (°C): 155
Melting point (°C): 46

https://us.vwr.com/stibo/web/std.lang.all/45/79/28934579.jpg

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR