2-Bromo-5-chlorobenzoic acid ≥98.0% (by GC, titration analysis)
Supplier: TCI America
B3112-5G
B3112-25G
TCB3112-25GEA
235.87
USD
TCB3112-25G
TCB3112-5G
2-Bromo-5-chlorobenzoic acid ≥98.0% (by GC, titration analysis)
2-Bromo-5-chlorobenzoic acid
CAS Number: 21739-93-5
MDL Number: MFCD00013982
Molecular Formula: C7H4BrClO2
Molecular Weight: 235.46
Purity/Analysis Method:
Form: Crystal
Melting point (°C): 155
Formula:
C₇H₄BrClO₂ Melting Pt: 155 °C |
MDL Number:
MFCD00013982 CAS Number: 21739-93-5 UN: 2811 ADR: 6.1,III |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...