Benzo[b]fluoranthene ≥98.0% (by GC)
Supplier: TCI America
B2982-100MG
TCB2982-100MGEA
151.24
USD
TCB2982-100MG
Benzo[b]fluoranthene ≥98.0% (by GC)
Benzo[b]fluoranthene
CAS Number: 205-99-2
MDL Number: MFCD00010582
Molecular Formula: C20H12
Molecular Weight: 252.32
Purity/Analysis Method:
Form: Crystal
Boiling point (°C): 481
Melting point (°C): 166
Formula:
C₂₀H₁₂ Boiling Pt: 481 °C Melting Pt: 166 °C |
MDL Number:
MFCD00010582 CAS Number: 205-99-2 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...