α-Bromo-2-chlorophenylacetic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: 2-Bromo-2-(2-chlorophenyl)acetic acid

B2950-25G
TCB2950-25GEA 31 USD
TCB2950-25G
α-Bromo-2-chlorophenylacetic acid ≥98.0% (by GC, titration analysis)
α-Bromo-2-chlorophenylacetic acid

CAS Number: 29270-30-2
MDL Number: MFCD03428513
Molecular Formula: C8H6BrClO2
Molecular Weight: 249.49
Purity/Analysis Method: 98.0% (GC,T)
Form: Crystal
Melting point (°C): 110

Formula: C₈H₆BrClO₂
MW: 249.49 g/mol
Melting Pt: 110 °C
MDL Number: MFCD03428513
CAS Number: 29270-30-2

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR