Bis(pyridine)iodonium tetrafluoroborate ≥95.0% (by titrimetric analysis)

Supplier: TCI America

B2539-1G
TCB2539-1GEA 210 USD
TCB2539-1G
Bis(pyridine)iodonium tetrafluoroborate ≥95.0% (by titrimetric analysis)
Bis(pyridine)iodonium tetrafluoroborate

CAS Number: 15656-28-7
MDL Number: MFCD03703393
Molecular Formula: C10H10BF4IN2
Molecular Weight: 371.91
Purity/Analysis Method: 95.0% (T)
Form: Crystal
Melting point (°C): 151
Storage Temperature: 0-10°C


Product Categories: Synthetic Organic Chemistry, Hypervalent Iodine Compounds, Oxidation, Halogenation
Other Product Names / Synonyms: Barluenga's Reagent

Formula: C₁₀H₁₀BF₄IN₂
Melting Pt: 151 °C
MDL Number: MFCD03703393
CAS Number: 15656-28-7
https://us.vwr.com/stibo/web/std.lang.all/41/78/28934178.jpg

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR