TCIAB2440

B2440-5G B2440-25G
TCB2440-25GEA 281.39 USD
TCB2440-25G TCB2440-5G
TCIAB2440
CAS Number: 202289-38-1
MDL Number: MFCD01321415
Molecular Formula: C6H14F3NO2S
Molecular Weight: 221.24
Purity/Analysis Method: 92.0% (T)
Form: Clear Liquid
Boiling point (°C): 71
Flash Point (°C): 96
Specific Gravity (20/20): 1.26
Storage Temperature: 0-10°C
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR