1,2-Benzisothiazol-3(2H)-one ≥98.0% (by GC)
Supplier: TCI America
Synonyms:
1,2-Benzisothiazol-3-one
B2430-25G
B2430-500G
TCB2430-500GEA
724.71
USD
TCB2430-500G
TCB2430-25G
1,2-Benzisothiazol-3(2H)-one ≥98.0% (by GC)
1,2-Benzisothiazol-3(2H)-one
CAS Number: 2634-33-5
MDL Number: MFCD00127753
Molecular Formula: C7H5NOS
Molecular Weight: 151.18
Purity/Analysis Method:
Form: Crystal
Melting point (°C): 158
Formula:
C₇H₅NOS Melting Pt: 158 °C |
MDL Number:
MFCD00044001 CAS Number: 2634-33-5 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...