3-Butyne-2-one ≥97.0% (by GC)

Supplier: TCI America

Synonyms: But-3-yn-2-one

B2393-5G
TCB2393-5GEA 269 USD
TCB2393-5G
3-Butyne-2-one ≥97.0% (by GC)
3-Butyne-2-one

CAS Number: 1423-60-5
MDL Number: MFCD00008775
Molecular Formula: C4H4O
Molecular Weight: 68.08
Purity/Analysis Method: 97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 86
Flash Point (°C): -1
Specific Gravity (20/20): 0.88
Storage Temperature: 0-10°C

Formula: C₄H₄O
MW: 68.08 g/mol
Boiling Pt: 86 °C
Density: 0.88 (20 °C)
Flash Pt: –1 °C
MDL Number: MFCD00008775
CAS Number: 1423-60-5
UN: 1992
ADR: 3,II
https://us.vwr.com/stibo/web/std.lang.all/41/30/28934130.jpg

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR