Benzylchlorodimethylsilane ≥98.0% (by GC)

Supplier: TCI America

B2334-5G
TCB2334-5GEA 168.49 USD
TCB2334-5G
Benzylchlorodimethylsilane ≥98.0% (by GC)
Benzylchlorodimethylsilane

CAS Number: 1833-31-4
MDL Number: MFCD00018089
Molecular Formula: C9H13ClSi
Molecular Weight: 184.74
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 211
Flash Point (°C): 73
Specific Gravity (20/20): 1.02

Formula: C₉H₁₃ClSi
Boiling Pt: 211 °C
Density: 1.02 (20 °C)
Flash Pt: 73 °C
MDL Number: MFCD00018089
CAS Number: 1833-31-4
https://us.vwr.com/stibo/web/std.lang.all/40/89/28934089.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR