9-Bromononanoic acid ≥97.0% (by titrimetric analysis)
Supplier: TCI America
B2323-1G
B2323-5G
TCB2323-1GEA
37.56
USD
TCB2323-1G
TCB2323-5G
9-Bromononanoic acid ≥97.0% (by titrimetric analysis)
9-Bromononanoic acid
CAS Number: 41059-02-3
MDL Number: MFCD00046564
Molecular Formula: C9H17BrO2
Molecular Weight: 237.14
Purity/Analysis Method:
Form: Crystal
Boiling point (°C): 174
Melting point (°C): 37
Storage Temperature: 0-10°C
Formula:
C₉H₁₇BrO₂ Boiling Pt: 174 °C Melting Pt: 37 °C |
MDL Number:
MFCD00046564 CAS Number: 41059-02-3 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...