4-Butylbenzoyl chloride ≥96.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: 4-Butylbenzene-1-carbonyl chloride

B2297-25G B2297-5G
TCB2297-25GEA 228 USD
TCB2297-25G TCB2297-5G
4-Butylbenzoyl chloride ≥96.0% (by GC, titration analysis)
4-Butylbenzoyl chloride

CAS Number: 28788-62-7
MDL Number: MFCD00000699
Molecular Formula: C11H13ClO
Molecular Weight: 196.67
Purity/Analysis Method: 96.0% (GC,T)
Form: Clear Liquid
Flash Point (°C): 113
Specific Gravity (20/20): 1.08

Formula: C₁₁H₁₃ClO
MW: 196.68 g/mol
Density: 1.08 (20 °C)
Flash Pt: 113 °C
MDL Number: MFCD00000699
CAS Number: 28788-62-7
UN: 3265
ADR: 8,II
https://us.vwr.com/stibo/web/std.lang.all/40/62/28934062.jpg

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR