(R)-(-)-1,1'-Binaphthyl-2,2'-diyl bis(trifluoromethanesulfonate) ≥96.0% (by HPLC)
Supplier: TCI America
Synonyms:
(R)-(-)-1,1'-Bi-2-naphthol Bis(trifluoromethanesulfonate)
B2230-1G
TCB2230-1GEA
47
USD
TCB2230-1G
(R)-(-)-1,1'-Binaphthyl-2,2'-diyl bis(trifluoromethanesulfonate) ≥96.0% (by HPLC)
(R)-(-)-1,1'-Binaphthyl-2,2'-diyl bis(trifluoromethanesulfonate)
CAS Number: 126613-06-7
MDL Number: MFCD00274615
Molecular Formula: C22H12F6O6S2
Molecular Weight: 550.44
Purity/Analysis Method:
Form: Crystal
Melting point (°C): 85
Specific rotation [a]20/D: -144 deg (C=1, THF)
Product Categories: Synthetic Organic Chemistry, Sulfonic Acid Derivatives, Asymmetric Synthesis
Other Product Names / Synonyms: (R)-(-)-1,1'-Bi-2-naphthol Bis(trifluoromethanesulfonate)
Formula:
C₂₂H₁₂F₆O₆S₂ MW: 550.44 g/mol Melting Pt: 85 °C |
MDL Number:
MFCD00274615 CAS Number: 126613-06-7 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...