TCIAB2209

B2209-1G
TCB2209-1GEA 287.22 USD
TCB2209-1G
TCIAB2209
CAS Number: 152227-36-6
Molecular Formula: C20H16Cl2N4Ru
Molecular Weight: 484.35
Purity/Analysis Method: 98.0% (T)
Form: Crystal
Lambda max.: 555 nm (DMF)

Other Product Names / Synonyms: cis-Bis(2,2'-bipyridine)dichlororuthenium(II), cis-Bis(2,2'-bipyridyl)ruthenium(II) Dichloride
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR