2,2,2-Trifluoroethyl butyrate ≥98.0% (by GC)
Supplier: TCI America
Synonyms:
Butyric acid 2,2,2-Trifluoroethyl ester
B2144-5G
B2144-25G
TCB2144-5GEA
82.22
USD
TCB2144-5G
TCB2144-25G
2,2,2-Trifluoroethyl butyrate ≥98.0% (by GC)
2,2,2-Trifluoroethyl butyrate
CAS Number: 371-27-7
MDL Number: MFCD00077609
Molecular Formula: C6H9F3O2
Molecular Weight: 170.13
Purity/Analysis Method:
Form: Clear Liquid
Boiling point (°C): 113
Flash Point (°C): 24
Specific Gravity (20/20): 1.14
Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: Butyric Acid 2,2,2-Trifluoroethyl Ester
Formula:
C₆H₉F₃O₂ MW: 170.1297 g/mol Boiling Pt: 113 °C Density: 1.14 (20 °C) Flash Pt: 24 °C |
MDL Number:
MFCD00077609 CAS Number: 371-27-7 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...