Bis[eta-(2,5-norbornadiene)]rhodium(I) tetrafluoroborate

Supplier: TCI America

Synonyms: Bis(norbornadiene)rhodium(I) tetrafluoroborateBis(norbornadiene)rhodium (I) tetrafluoroborate

B2091-100MG
TCB2091-100MGEA 106.19 USD
TCB2091-100MG
Bis[eta-(2,5-norbornadiene)]rhodium(I) tetrafluoroborate
Bis[eta-(2,5-norbornadiene)]rhodium(I) tetrafluoroborate

CAS Number: 36620-11-8
MDL Number: MFCD00671775
Molecular Formula: C14H16BF4Rh
Molecular Weight: 373.99
Form: Crystal
Color: Yellow
Melting point (°C): 140

Formula: C₁₄H₁₆BF₄Rh
MW: 365,92 g/mol
Melting Pt: 140 °C
MDL Number: MFCD00671775
CAS Number: 36620-11-8
https://us.vwr.com/stibo/web/std.lang.all/38/59/28933859.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR