2,4-Bis(chloromethyl)-1,3,5-trimethylbenzene ≥98.0% (by GC)

Supplier: TCI America

Synonyms: 2,4-Bis(chloromethyl)mesitylene

B1994-25G
TCB1994-25GEA 58 USD
TCB1994-25G
2,4-Bis(chloromethyl)-1,3,5-trimethylbenzene ≥98.0% (by GC)
2,4-Bis(chloromethyl)-1,3,5-trimethylbenzene

CAS Number: 1585-17-7
MDL Number: MFCD00013681
Molecular Formula: C11H14Cl2
Molecular Weight: 217.13
Purity/Analysis Method: 98.0% (GC)
Form: Crystal
Boiling point (°C): 180
Melting point (°C): 107


Other Product Names / Synonyms: 2,4-Bis(chloromethyl)mesitylene

Formula: C₁₁H₁₄Cl₂
Boiling Pt: 180 °C
Melting Pt: 107 °C
MDL Number: MFCD00013681
CAS Number: 1585-17-7
UN: 3261
ADR: 8,III
https://us.vwr.com/stibo/web/std.lang.all/37/53/28933753.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR