TBTU (O-(1H-benzotriazol-1-yl)-N,N,N',N'-tetramethyluronium tetrafluoroborate) ≥98.0% (by HPLC)
Supplier: TCI America
Synonyms:
TBTU
B1658-5G
B1658-25G
TCB1658-5GEA
44
USD
TCB1658-5G
TCB1658-25G
TBTU (O-(1H-benzotriazol-1-yl)-N,N,N',N'-tetramethyluronium tetrafluoroborate) ≥98.0% (by HPLC)
TBTU (O-(1H-Benzotriazol-1-yl)-N,N,N',N'-tetramethyluronium tetrafluoroborate)
[Coupling Reagent for Peptide]
CAS Number: 125700-67-6
MDL Number: MFCD00077413
Molecular Formula: C11H16BF4N5O
Molecular Weight: 321.09
Purity/Analysis Method:
Form: Crystal
Melting point (°C): 205
Product Categories: Synthetic Organic Chemistry, Condensation & Active Esterification, Peptide Synthesis, Peptide Synthesis
Other Product Names / Synonyms: TBTU, N,N,N',N'-Tetramethyl-O-(benzotriazol-1-yl)uronium Tetrafluoroborate
Formula:
C₁₁H₁₆BF₄N₅O MW: 321.1 g/mol Melting Pt: 205 °C |
MDL Number:
MFCD00077413 CAS Number: 125700-67-6 |
![]() |
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...