Trimethylsilyl bromoacetate ≥97.0% (by GC)

Supplier: TCI America

B1578-5G B1578-25G
TCB1578-5GEA 52 USD
TCB1578-5G TCB1578-25G
Trimethylsilyl bromoacetate ≥97.0% (by GC)
Trimethylsilyl bromoacetate

CAS Number: 18291-80-0
MDL Number: MFCD00000187
Molecular Formula: C5H11BrO2Si
Molecular Weight: 211.13
Purity/Analysis Method: 97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 83
Flash Point (°C): 28
Specific Gravity (20/20): 1.30


Other Product Names / Synonyms: Bromoacetic Acid Trimethylsilyl Ester

Formula: C₅H₁₁BrO₂Si
Boiling Pt: 83 °C
Density: 1.30 (20 °C)
Flash Pt: 28 °C
MDL Number: MFCD00000187
CAS Number: 18291-80-0
UN: 2920
ADR: 8,II
https://us.vwr.com/stibo/web/std.lang.all/33/44/28933344.jpg

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR