(3-Bromo-4-methoxyphenyl)acetonitrile ≥98.0% (by GC)

Supplier: TCI America

Synonyms: 2-(3-Bromo-4-methoxyphenyl)acetonitrile

B1455-25G
TCB1455-25GEA 248.49 USD
TCB1455-25G
(3-Bromo-4-methoxyphenyl)acetonitrile ≥98.0% (by GC)
(3-Bromo-4-methoxyphenyl)acetonitrile

CAS Number: 772-59-8
MDL Number: MFCD00016391
Molecular Formula: C9H8BrNO
Molecular Weight: 226.07
Purity/Analysis Method: 98.0% (GC)
Form: Crystal
Boiling point (°C): 178
Melting point (°C): 56


Other Product Names / Synonyms: 3-Bromo-4-methoxyphenylacetonitrile

Formula: C₉H₈BrNO
MW: 226.08 g/mol
Boiling Pt: 178 °C
Melting Pt: 56 °C
UN: 3439
ADR: 6.1,III
https://us.vwr.com/stibo/web/std.lang.all/32/45/28933245.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR