Bis(2-methoxyethyl)amine ≥98.0% (by GC, titration analysis)

Supplier: TCI America

B1383-25ML
TCB1383-25MLEA 100.24 USD
TCB1383-25ML
Bis(2-methoxyethyl)amine ≥98.0% (by GC, titration analysis)
Bis(2-methoxyethyl)amine

CAS Number: 111-95-5
MDL Number: MFCD00025906
Molecular Formula: C6H15NO2
Molecular Weight: 133.19
Purity/Analysis Method: 98.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 172
Flash Point (°C): 58
Specific Gravity (20/20): 0.91


Other Product Names / Synonyms: 2,2'-Dimethoxydiethylamine

Formula: C₆H₁₅NO₂
MW: 133.1888 g/mol
Boiling Pt: 172 °C
Density: 0.91 (20 °C)
Flash Pt: 58 °C
MDL Number: MFCD00025906
CAS Number: 111-95-5
https://us.vwr.com/stibo/web/std.lang.all/31/88/28933188.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR