Borane-tert-butylamine complex ≥95.0% (by titrimetric analysis)
Supplier: TCI America
Synonyms:
Boron hydride-tert-butylamine
B1264-100G
B1264-25G
TCB1264-100GEA
387.73
USD
TCB1264-100G
TCB1264-25G
Borane-tert-butylamine complex ≥95.0% (by titrimetric analysis)
Borane tert-butylamine complex
CAS Number: 7337-45-3
MDL Number: MFCD00075635
Molecular Formula: C4H14BN
Molecular Weight: 86.97
Purity/Analysis Method:
Form: Crystal
Melting point (°C): 99
Product Categories: Synthetic Organic Chemistry, Reduction
Other Product Names / Synonyms: tert-Butylamine Borane, TBAB
Formula:
C₄H₁₄BN Melting Pt: 99 °C |
MDL Number:
MFCD00075635 CAS Number: 7337-45-3 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...