2-Bromohexane ≥70.0% (by GC) (contains 3-Bromohexane) stabilized

Supplier: TCI America

B1235-25G
TCB1235-25GEA 173.49 USD
TCB1235-25G
2-Bromohexane ≥70.0% (by GC) (contains 3-Bromohexane) stabilized
2-Bromohexane

(stabilized with Copper chip)
CAS Number: 3377-86-4
MDL Number: MFCD00039183
Molecular Formula: C6H13Br
Molecular Weight: 165.07
Purity/Analysis Method: 70.0% (GC)
Form: Clear Liquid
Flash Point (°C): 47
Specific Gravity (20/20): 1.17


Other Product Names / Synonyms: 2-Hexyl Bromide

Formula: H₃CBrCH(CH₂)₃CH₃
MW: 165.07 g/mol
Density: 1.17 (20 °C)
Flash Pt: 47 °C
MDL Number: MFCD00039183
CAS Number: 3377-86-4
UN: 1993
ADR: 3,III
https://us.vwr.com/stibo/web/std.lang.all/30/68/28933068.jpg

Order Now

Specification Test Results

Purity/Analysis Method 70.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR