Bis(methylthio)methane ≥98.0% (by GC)

Supplier: TCI America

B1204-25ML B1204-5ML
TCB1204-25MLEA 85 USD
TCB1204-25ML TCB1204-5ML
Bis(methylthio)methane ≥98.0% (by GC)
Bis(methylthio)methane

CAS Number: 1618-26-4
MDL Number: MFCD00008564
Molecular Formula: C3H8S2
Molecular Weight: 108.22
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 152
Flash Point (°C): 44
Specific Gravity (20/20): 1.06


Other Product Names / Synonyms: Bis(methylmercapto)methane, Formaldehyde Dimethyl Mercaptal, Methylenebis(methyl Sulfide)

Formula: C₃H₈S₂
Boiling Pt: 152 °C
Density: 1.06 (20 °C)
Flash Pt: 44 °C
MDL Number: MFCD00008564
CAS Number: 1618-26-4
https://us.vwr.com/stibo/web/std.lang.all/30/40/28933040.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR