N-(tert-Butyldimethylsilyl)-N-methyltrifluoroacetamide (MTBSTFA) ≥95.0% (by GC)
Supplier: TCI America
Synonyms:
N-(tert-Butyldimethylsilyl)-2,2,2-trifluoro-N-methylacetamide
, MBDSTFA
B1150-10G
B1150-1G
TCB1150-10GEA
137.03
USD
TCB1150-10G
TCB1150-1G
N-(tert-Butyldimethylsilyl)-N-methyltrifluoroacetamide (MTBSTFA) ≥95.0% (by GC)
N-(tert-Butyldimethylsilyl)-N-methyltrifluoroacetamide (MTBSTFA)
CAS Number: 77377-52-7
MDL Number: MFCD00009671
Molecular Formula: C9H18F3NOSi
Molecular Weight: 241.33
Purity/Analysis Method:
Form: Clear Liquid
Boiling point (°C): 170
Flash Point (°C): 52
Specific Gravity (20/20): 1.03
Product Categories: Analytical Science, Silylation (GC Derivatizing Reagents)
Other Product Names / Synonyms: N-Methyl-N-(tert-butyldimethylsilyl)trifluoroacetamide, MTBSTFA
Formula:
C₉H₁₈F₃NOSi MW: 241.33 g/mol Boiling Pt: 170 °C Density: 1.03 (20 °C) Flash Pt: 52 °C |
MDL Number:
MFCD00009671 CAS Number: 77377-52-7 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...