1,3-Bis(trimethylsilyl)urea ≥98.0% (by total nitrogen basis)
Supplier: TCI America
Synonyms:
N,N'-Bis(trimethylsilyl)urea
B1103-25G
TCB1103-25GEA
84.25
USD
TCB1103-25G
1,3-Bis(trimethylsilyl)urea ≥98.0% (by total nitrogen basis)
1,3-Bis(trimethylsilyl)urea
CAS Number: 18297-63-7
MDL Number: MFCD00008260
Molecular Formula: C7H20N2OSi2
Molecular Weight: 204.42
Purity/Analysis Method:
Form: Crystal
Product Categories: Synthetic Organic Chemistry, Ureas (Multi-component Reaction), Silicon Compounds (for Synthesis)
Other Product Names / Synonyms: BSU
Formula:
C₇H₂₀N₂OSi₂ |
MDL Number:
MFCD00008260 CAS Number: 18297-63-7 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...