9-Bromofluorene ≥98.0% (by titrimetric analysis)

Supplier: TCI America

B1046-25G
TCB1046-25GEA 131 USD
TCB1046-25G
9-Bromofluorene ≥98.0% (by titrimetric analysis)
9-Bromofluorene

CAS Number: 1940-57-4
MDL Number: MFCD00001133
Molecular Formula: C13H9Br
Molecular Weight: 245.12
Purity/Analysis Method: 98.0% (T)
Form: Crystal
Melting point (°C): 104


Other Product Names / Synonyms: 9-Fluorenyl Bromide

Formula: C₁₃H₉Br
MW: 245.12 g/mol
Melting Pt: 104 °C
MDL Number: MFCD00001133
CAS Number: 1940-57-4
UN: 3261
ADR: 8,II
https://us.vwr.com/stibo/web/std.lang.all/29/09/28932909.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR