2-Butyne ≥97.0% (by GC)

Supplier: TCI America

B0951-5ML
TCB0951-5MLEA 69.02 USD
TCB0951-5ML
2-Butyne ≥97.0% (by GC)
2-Butyne

CAS Number: 503-17-3
MDL Number: MFCD00009275
Molecular Formula: C4H6
Molecular Weight: 54.09
Purity/Analysis Method: 97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 27
Flash Point (°C): -20
Specific Gravity (20/20): 0.69
Storage Temperature: 0-10°C


Other Product Names / Synonyms: Dimethylacetylene

Formula: C₄H₆
MW: 54.09 g/mol
Boiling Pt: 27 °C
Density: 0.69 (20 °C)
Flash Pt: –20 °C
MDL Number: MFCD00009275
CAS Number: 503-17-3
UN: 3295
ADR: 3,I
https://us.vwr.com/stibo/web/std.lang.all/28/30/28932830.jpg

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR