4-Bromo-1-butene ≥97.0% (by GC)

Supplier: TCI America

B0920-10G B0920-25G B0920-250G
TCB0920-250GEA 465.69 USD
TCB0920-250G TCB0920-10G TCB0920-25G
4-Bromo-1-butene ≥97.0% (by GC)
4-Bromo-1-butene

CAS Number: 5162-44-7
MDL Number: MFCD00000258
Molecular Formula: C4H7Br
Molecular Weight: 135.00
Purity/Analysis Method: 97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 99
Flash Point (°C): 1
Specific Gravity (20/20): 1.34


Other Product Names / Synonyms: 3-Butenyl Bromide

Formula: C₄H₇Br
MW: 135 g/mol
Boiling Pt: 99 °C
Density: 1.34 (20 °C)
Flash Pt: 1 °C
MDL Number: MFCD00000258
CAS Number: 5162-44-7
UN: 1992
ADR: 3,II
https://us.vwr.com/stibo/web/std.lang.all/28/01/28932801.jpg

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR