TCIAB0825

B0825-5G
TCB0825-5GEA 124.68 USD
TCB0825-5G
TCIAB0825
CAS Number: 4712-38-3
MDL Number: MFCD00002965
Molecular Formula: C4H9DO
Molecular Weight: 75.13
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 118
Melting point (°C): -90
Flash Point (°C): 34

Product Categories: Analytical Science, Deuterated Compounds for NMR
Other Product Names / Synonyms: Monodeuterobutyl Alcohol
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR