Butyl ethyl ether ≥99.0% (by GC)

Supplier: TCI America

B0776-25ML
TCB0776-25MLEA 59.06 USD
TCB0776-25ML
Butyl ethyl ether ≥99.0% (by GC)
Butyl ethyl ether

CAS Number: 628-81-9
MDL Number: MFCD00009455
Molecular Formula: C6H14O
Molecular Weight: 102.18
Purity/Analysis Method: 99.0% (GC)
Form: Clear Liquid
Boiling point (°C): 92
Flash Point (°C): 4
Specific Gravity (20/20): 0.75

Formula: C₆H₁₄O
Boiling Pt: 92 °C
Density: 0.75 (20 °C)
Flash Pt: 4 °C
MDL Number: MFCD00009455
CAS Number: 628-81-9
UN: 1179
ADR: 3,II
https://us.vwr.com/stibo/web/std.lang.all/26/99/28932699.jpg

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR