4-tert-Butylphenol ≥98.0% (by GC), zone refined: number of passes 19
Supplier: TCI America
B0735-1SAMPLE
TCB0735-ZONEEA
287
USD
TCB0735-ZONE
4-tert-Butylphenol ≥98.0% (by GC), zone refined: number of passes 19
4-tert-Butylphenol
CAS Number: 98-54-4
MDL Number: MFCD00002367
Molecular Formula: C10H14O
Molecular Weight: 150.22
Purity/Analysis Method:
Form: Crystal
Boiling point (°C): 238
Melting point (°C): 101
Flash Point (°C): 113
Formula:
C₁₀H₁₄O MW: 150.22 g/mol Boiling Pt: 238 °C Melting Pt: 101 °C Flash Pt: 113 °C |
MDL Number:
MFCD00002367 CAS Number: 98-54-4 UN: 2430 ADR: 8,III |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...