(±)-2-Bromoisovaleric acid ≥90.0% (by GC)

Supplier: TCI America

Synonyms: 2-Bromo-3-methylbutyricacid2-Bromo-3-methylbutyric acid

B0668-25G
TCB0668-25GEA 238 USD
TCB0668-25G
(±)-2-Bromoisovaleric acid ≥90.0% (by GC)
(±)-2-Bromoisovaleric acid

CAS Number: 565-74-2
MDL Number: MFCD00004210
Molecular Formula: C5H9BrO2
Molecular Weight: 181.03
Purity/Analysis Method: 90.0% (GC)
Form: Crystal
Boiling point (°C): 230
Flash Point (°C): 107
Freezing point (°C): 36


Other Product Names / Synonyms: 2-Bromoisovaleric Acid

Formula: C₅H₉BrO₂
MW: 181.03 g/mol
Boiling Pt: 230 °C
Flash Pt: 107 °C
MDL Number: MFCD00004210
CAS Number: 565-74-2
UN: 3261
ADR: 8,II
https://us.vwr.com/stibo/web/std.lang.all/26/04/28932604.jpg

Order Now

Specification Test Results

Purity/Analysis Method 90.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR