Benzyltrimethylammonium hydroxide 10% in water

Supplier: TCI America

Synonyms: Benzyltrimethylammonium HydroxideN,N,N-Trimethyl-1-phenylmethanaminium Hydroxide

B0428-25ML
TCB0428-25MLEA 42 USD
TCB0428-25ML
Benzyltrimethylammonium hydroxide 10% in water
Benzyltrimethylammonium hydroxide

CAS Number: 100-85-6
MDL Number: MFCD00008281
Molecular Formula: C10H17NO
Molecular Weight: 167.25
Form: Clear Liquid
Color: Colorless
Specific Gravity (20/20): 1.02

Formula: C₁₀H₁₇NO
MW: 167.25 g/mol
Density: 1.02 (20 °C)
UN: 3286
ADR: 3,I
https://us.vwr.com/stibo/web/std.lang.all/23/98/28932398.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR