Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Butyltrichlorosilane ≥98.0% (by GC)

Supplier: TCI America
Synonyms: Trichloro(butyl)silanen-Butyltrichlorosilane

B0393-25G
TCB0393-25G
Butyltrichlorosilane ≥98.0% (by GC)
CAS Number: 7521-80-4
MDL Number: MFCD00013605
Molecular Formula: C4H9Cl3Si
Molecular Weight: 191.55
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 150
Flash Point (°C): 54
Specific Gravity (20/20): 1.17

Product Categories: Materials Chemistry, Silane Coupling Agents
Other Product Names / Synonyms: Trichlorobutylsilane


Formula: C₄H₉Cl₃Si
MW: 191,56 g/mol
Boiling Pt: 132 to 134 °C (1013 hPa)
Density: 1.16 g/cm³ (20 °C)
Flash Pt: 54 °C
MDL Number: MFCD00013605
CAS Number: 7521-80-4
EINECS: 231-381-3
UN: 1747
ADR: 8,II
Order Now
Specification Test Results
Purity/Analysis Method >98.0% (GC)
Boiling point (°C) 150
Flash Point (°C) 54
Specific Gravity (20/20) 1.17


 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty