3-(4-Nitrophenyl)-5-methyl-2-phenyltetrazolium chloride ≥98.0% (by HPLC)

Supplier: TCI America

Synonyms: 2-Phenyl-3-(4-nitrophenyl)-5-methyltetrazolium Chloride

B0311-100MG
TCB0311-100MGEA 100 USD
TCB0311-100MG
3-(4-Nitrophenyl)-5-methyl-2-phenyltetrazolium chloride ≥98.0% (by HPLC)
3-(4-Nitrophenyl)-5-methyl-2-phenyltetrazolium chloride

MDL Number: MFCD00060011
Molecular Formula: C14H12ClN5O2
Molecular Weight: 317.73
Purity/Analysis Method: 98.0% (HPLC)
Form: Crystal


Other Product Names / Synonyms: 2-Phenyl-3-(4-nitrophenyl)-5-methyltetrazolium Chloride

https://us.vwr.com/stibo/web/std.lang.all/23/52/28932352.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR