Biphenyl (PCB No. 0) ≥99.0% (by GC), zone refined: number of passes 24

Supplier: TCI America

Synonyms: Phenylbenzene Diphenyl PCB No. 0 0-PCB

B0224-1SAMPLE
TCB0224-ZONEEA 221 USD
TCB0224-ZONE
Biphenyl (PCB No. 0) ≥99.0% (by GC), zone refined: number of passes 24
Biphenyl (PCB No. 0)

CAS Number: 92-52-4
MDL Number: MFCD00003054
Molecular Formula: C12H10
Molecular Weight: 154.21
Purity/Analysis Method: 99.0% (GC)
Form: Crystal
Boiling point (°C): 256
Melting point (°C): 70
Flash Point (°C): 113

Formula: C₁₂H₁₀
MW: 154.21 g/mol
Boiling Pt: 256 °C
Melting Pt: 70 °C
Flash Pt: 113 °C
MDL Number: MFCD00003054
CAS Number: 92-52-4
UN: 3077
ADR: 9,III
https://us.vwr.com/stibo/web/std.lang.all/23/04/28932304.jpg

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR