α-Bromo-p-xylene ≥98.0% (by GC)
Supplier: TCI America
Synonyms:
alpha-Bromo-p-xylene
, 4-(Bromomethyl)toluene
, 4-Methylbenzyl bromide
B0138-25G
TCB0138-25GEA
140.07
USD
TCB0138-25G
α-Bromo-p-xylene ≥98.0% (by GC)
α-Bromo-p-xylene
CAS Number: 104-81-4
MDL Number: MFCD00000181
Molecular Formula: C8H9Br
Molecular Weight: 185.06
Purity/Analysis Method:
Form: Crystal
Boiling point (°C): 220
Flash Point (°C): 97
Freezing point (°C): 35
Other Product Names / Synonyms: 4-Methylbenzyl Bromide
Formula:
C₈H₉Br MW: 185.06 g/mol Boiling Pt: 220 °C Flash Pt: 97 °C |
MDL Number:
MFCD00000181 CAS Number: 104-81-4 UN: 3417 ADR: 6.1,II |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...