Phenyl isothiocyanate ≥99.0% (by GC)

Supplier: TCI America

A5513-5ML
TCA5513-5MLEA 42.63 USD
TCA5513-5ML
Phenyl isothiocyanate ≥99.0% (by GC)
Phenyl isothiocyanate

[for HPLC Labeling]
CAS Number: 103-72-0
MDL Number: MFCD00004798
Molecular Formula: C7H5NS
Molecular Weight: 135.18
Purity/Analysis Method: 99.0% (GC)
Form: Clear Liquid
Boiling point (°C): 71
Flash Point (°C): 88
Freezing point (°C): -21
Specific Gravity (20/20): 1.14
Storage Temperature: 0-10°C


Product Categories: Analytical Science, UV Detection (HPLC Labeling Reagents)
Other Product Names / Synonyms: Isothiocyanic Acid Phenyl Ester

Formula: C₇H₅NS
MW: 135.19 g/mol
Boiling Pt: 71 °C
Density: 1.14 (20 °C)
Flash Pt: 88 °C
MDL Number: MFCD00004798
CAS Number: 103-72-0
UN: 2927
ADR: 6.1,II
https://us.vwr.com/stibo/web/std.lang.all/21/15/28932115.jpg

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR