ω,4-Dibromoacetophenone ≥99.0% (by titrimetric analysis)
Supplier: TCI America
Synonyms:
2,4'-Dibromoacetophenone
[for HPLC Labeling]
CAS Number: 99-73-0
MDL Number: MFCD00000200
Molecular Formula: C8H6Br2O
Molecular Weight: 277.94
Purity/Analysis Method:
Form: Crystal
Melting point (°C): 111
Storage Temperature: 0-10°C
Product Categories: Analytical Science, UV Detection (HPLC Labeling Reagents)
Other Product Names / Synonyms: 2,4'-Dibromoacetophenone
Formula:
C₈H₆Br₂O MW: 277.94 g/mol Melting Pt: 111 °C |
MDL Number:
MFCD00000200 CAS Number: 99-73-0 UN: 3261 ADR: 8,II |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...