Crystal violet nonahydrate ≥97.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: Basic Violet 3

A5104-1G
TCA5104-1GEA 56.81 USD
TCA5104-1G
Crystal violet nonahydrate ≥97.0% (by titrimetric analysis)
Crystal violet nonahydrate

[Ion association reagent for spectrophotometric analysis]
CAS Number: 60662-33-1
MDL Number: MFCD00011750
Molecular Formula: C25H30ClN3
Molecular Weight: 407.99
Purity/Analysis Method: 97.0% (T)
Form: Crystal
Melting point (°C): 211


Product Categories: Analytical Science, Ion Association Reagents
Other Product Names / Synonyms: Basic Violet 3

Formula: C₂₅H₃₀ClN₃
MW: 407.99 g/mol
Melting Pt: 211 °C
MDL Number: MFCD00011750
CAS Number: 60662-33-1
https://us.vwr.com/stibo/web/std.lang.all/20/77/28932077.jpg

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR