TCIAA2794

A2794-1G A2794-5G
TCA2794-5GEA 409.67 USD
TCA2794-5G TCA2794-1G
TCIAA2794
CAS Number: 6777-00-0
MDL Number: MFCD09908098
Molecular Formula: C6H10O2
Molecular Weight: 114.14
Purity/Analysis Method: 97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 64
Specific Gravity (20/20): 0.98
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR