N-Acetyltryptamine ≥98.0% (by HPLC)

Supplier: TCI America

Synonyms: 3-(2-Acetamidoethyl)indoleN-(2-(1H-Indol-3-yl)ethyl)acetamide

A2685-100MG A2685-25MG
TCA2685-25MGEA 166.41 USD
TCA2685-25MG TCA2685-100MG
N-Acetyltryptamine ≥98.0% (by HPLC)
N-Acetyltryptamine

CAS Number: 1016-47-3
MDL Number: MFCD00209910
Molecular Formula: C12H14N2O
Molecular Weight: 202.26
Purity/Analysis Method: 98.0% (HPLC)
Form: Crystal
Boiling point (°C): 200
Melting point (°C): 76
Lambda max.: 290 nm (MeOH)


Other Product Names / Synonyms: 3-(2-Acetamidoethyl)indole

https://us.vwr.com/stibo/web/std.lang.all/16/66/28931666.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR