Agomelatine ≥98.0% (by HPLC, total nitrogen)

Supplier: TCI America

Synonyms: N-[2-(7-Methoxy-1-naphthalenyl)ethyl]acetamide

A2606-1G
TCA2606-1GEA 360.22 USD
TCA2606-1G
Agomelatine ≥98.0% (by HPLC, total nitrogen)
Agomelatine

CAS Number: 138112-76-2
MDL Number: MFCD00916659
Molecular Formula: C15H17NO2
Molecular Weight: 243.31
Purity/Analysis Method: 98.0% (HPLC,N)
Form: Crystal
Melting point (°C): 110
Storage Temperature: 0-10°C


Product Categories: Bioscience, Neurotransmitter Agent Ingredients
Other Product Names / Synonyms: N-[2-(7-Methoxy-1-naphthalenyl)ethyl]acetamide

Formula: C₁₅H₁₇NO₂
MW: 243.31 g/mol
Melting Pt: 110 °C
MDL Number: MFCD00916659
CAS Number: 138112-76-2
https://us.vwr.com/stibo/web/std.lang.all/16/10/28931610.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,N)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR