Alfuzosin hydrochloride ≥98.0% (by HPLC)

Supplier: TCI America

Synonyms: N-[3-[(4-Amino-6,7-dimethoxy-2-quinazolinyl)(methyl)amino]propyl]tetrahydro-2-furancarboxamide Hydrochloride4-Amino-6,7-dimethoxy-2-[methyl[3-[(tetrahydro-2-furoyl)amino]propyl]amino]quinazoline Hydrochloride

A2591-100MG A2591-1G
TCA2591-100MGEA 61.41 USD
TCA2591-100MG TCA2591-1G
Alfuzosin hydrochloride ≥98.0% (by HPLC)
Alfuzosin hydrochloride

CAS Number: 81403-68-1
MDL Number: MFCD00879135
Molecular Formula: C19H27N5O4
Molecular Weight: 425.91
Purity/Analysis Method: 98.0% (HPLC)
Form: Crystal
Melting point (°C): 229


Product Categories: Bioscience, Other Pharmaceutical Ingredients
Other Product Names / Synonyms: N-[3-[(4-Amino-6,7-dimethoxy-2-quinazolinyl)(methyl)amino]propyl]tetrahydro-2-furancarboxamide Hydrochloride, 4-Amino-6,7-dimethoxy-2-[methyl[3-[(tetrahydro-2-furoyl)amino]propyl]amino]quinazoline Hydrochloride

Formula: C₁₉H₂₇N₅O₄
MW: 425.91 g/mol
Melting Pt: 229 °C
MDL Number: MFCD00879135
CAS Number: 81403-68-1
https://us.vwr.com/stibo/web/std.lang.all/07/76/28930776.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR