5-Amino-2-chloro-3-methylpyridine ≥97.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: 6-Chloro-5-methylpyridin-3-amine5-Amino-2-chloro-3-picoline

A2564-5G
TCA2564-5GEA 88.31 USD
TCA2564-5G
5-Amino-2-chloro-3-methylpyridine ≥97.0% (by GC, titration analysis)
5-Amino-2-chloro-3-methylpyridine

CAS Number: 38186-82-2
MDL Number: MFCD03095087
Molecular Formula: C6H7ClN2
Molecular Weight: 142.59
Purity/Analysis Method: 97.0% (GC,T)
Form: Crystal
Melting point (°C): 93


Other Product Names / Synonyms: 5-Amino-2-chloro-3-picoline

Formula: C₆H₇ClN₂
MW: 142.5900 g/mol
Melting Pt: 93 °C
MDL Number: MFCD03095087
CAS Number: 38186-82-2
https://us.vwr.com/stibo/web/std.lang.all/07/64/28930764.jpg

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR