5-Amino-2-methylbenzenesulfonamide ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
A2552-5G
A2552-25G
TCA2552-5GEA
26.36
USD
TCA2552-5G
TCA2552-25G
5-Amino-2-methylbenzenesulfonamide ≥98.0% (by HPLC, titration analysis)
5-Amino-2-methylbenzenesulphonamide
CAS Number: 6973-09-7
MDL Number: MFCD06681071
Molecular Formula: C7H10N2O2S
Molecular Weight: 186.23
Purity/Analysis Method:
Form: Crystal
Melting point (°C): 169
Formula:
C₇H₁₀N₂O₂S MW: 186.23 g/mol Melting Pt: 169 °C |
MDL Number:
MFCD06681071 CAS Number: 6973-09-7 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...