N-(3-Azidopropyl)biotinamide ≥95.0% (by HPLC)

Supplier: TCI America

A2524-100MG
TCA2524-100MGEA 508.52 USD
TCA2524-100MG
N-(3-Azidopropyl)biotinamide ≥95.0% (by HPLC)
N-(3-Azidopropyl)biotinamide

CAS Number: 908007-17-0
MDL Number: MFCD20134144
Molecular Formula: C13H22N6O2S
Molecular Weight: 326.42
Purity/Analysis Method: 95.0% (HPLC)
Form: Crystal
Melting point (°C): 168

Formula: C₁₃H₂₂N₆O₂S
Melting Pt: 168 °C
MDL Number: MFCD20134144
CAS Number: 908007-17-0
https://us.vwr.com/stibo/web/std.lang.all/07/28/28930728.jpg

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (HPLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR