3-(Aminomethyl)piperidine ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: Piperidin-3-ylmethanamine

A2518-5G
TCA2518-5GEA 93.79 USD
TCA2518-5G
3-(Aminomethyl)piperidine ≥98.0% (by GC, titration analysis)
3-(Aminomethyl)piperidine

CAS Number: 23099-21-0
MDL Number: MFCD03427344
Molecular Formula: C6H14N2
Molecular Weight: 114.19
Purity/Analysis Method: 98.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 89
Flash Point (°C): 77
Specific Gravity (20/20): 0.96

Formula: C₆H₁₄N₂
MW: 114.19 g/mol
Boiling Pt: 89 °C
Density: 0.96 (20 °C)
Flash Pt: 77 °C
MDL Number: MFCD03427344
CAS Number: 23099-21-0
https://us.vwr.com/stibo/web/std.lang.all/07/23/28930723.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR