Acetophenone-O-benzoyloxime ≥98.0% (by HPLC, total nitrogen)

Supplier: TCI America

A2502-1G A2502-5G
TCA2502-5GEA 456.26 USD
TCA2502-5G TCA2502-1G
Acetophenone-O-benzoyloxime ≥98.0% (by HPLC, total nitrogen)
Acetophenone-O-benzoyloxime

CAS Number: 26060-56-0
Molecular Formula: C15H13NO2
Molecular Weight: 239.27
Purity/Analysis Method: 98.0% (HPLC,N)
Form: Crystal
Melting point (°C): 100

Formula: C₁₅H₁₃NO₂
MW: 239.27 g/mol
Melting Pt: 100 °C
CAS Number: 26060-56-0
https://us.vwr.com/stibo/web/std.lang.all/07/12/28930712.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,N)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR