14-Azido-3,6,9,12-tetraoxatetradecanol ≥95.0% (by GC)

Supplier: TCI America

Synonyms: PEG5-Azide

A2500-100MG
TCA2500-100MGEA 125.83 USD
TCA2500-100MG
14-Azido-3,6,9,12-tetraoxatetradecanol ≥95.0% (by GC)
14-Azido-3,6,9,12-tetraoxatetradecanol

CAS Number: 86770-68-5
MDL Number: MFCD20134131
Molecular Formula: C10H21N3O5
Molecular Weight: 263.29
Purity/Analysis Method: 95.0% (GC)
Form: Clear Liquid
Flash Point (°C): 100
Storage Temperature: 0-10°C


Product Categories: Synthetic Organic Chemistry, Azide Compounds (Click Chemistry), Bioscience, Crosslinkers, PEGylation Reagents & PEG Spacers
Other Product Names / Synonyms: PEG5-Azide

Formula: C₁₀H₂₁N₃O₅
MW: 263.29 g/mol
Flash Pt: 100 °C
MDL Number: MFCD20134131
CAS Number: 86770-68-5
https://us.vwr.com/stibo/web/std.lang.all/07/10/28930710.jpg

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR